About this item
VIAFLO 96 is a handheld 96-channel pipetting solution designed to fit on any lab bench. The system works with 96-channel heads only. VIAFLO 384 is an enhanced version of the VIAFLO 96 pipette. It features the same footprint and intuitive operation but allows working with both 96- and 384-channel heads.
- Bench top sized, require little space and fit into laminar flow hoods
- Pipetting heads can quickly be changed to adapt VIAFLO 96/384 to the application's volume range
- Ten predefined pipetting modes, optional set up of up to 20 personalized custom programs
- GripTips together with the new tip fitting interface form a integral system, preventing effectively tips from falling off and guaranteeing a perfect sealing to achieve best pipetting results
- Touch wheel controlled graphical user interface
- Two plate holders on the instrument base accept tip boxes, microplates or reagent reservoirs
These two systems close the gap between traditional manual pipettes and robotic systems. The user concept resembles traditional manual pipetting. VIAFLO 96/384 is hand guided but movements are assisted by motors. This enables an effortless and precise workflow. Pushing in one direction steers the pipetting head that way, making VIAFLO 96/384 incredibly intuitive to operate. Everyone in the laboratory is able to use the instrument without special training.
The systems are ideal for fast and convenient sample transfers, either from plate to plate, from reservoir to plate or within plates.
GripTips: GripTips are offered in non sterile, sterile and sterile filtered configurations to satisfy all application demands. Filtered GripTips reliably protect the pipetting head against contaminations caused by aerosols. Larger volume GripTips (300, 1250 μl) are delivered in boxes of 96 tips. The smaller volume GripTips (12,5, 125 μl) are filled in a space-efficient 384-fold grid in XYZ boxes. These boxes house 4 times as many tips and feature the standard SBS-footprint.
Specifications
- Catalog No:
- INTL6031
- INTL6001
- 型号:
- VIAFLO 384
- VIAFLO 96
- 移液管通道数量:
- 96, 384
- 96
- 电力供应:
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 系统宽度×深度×高度:
- 420×300×540 mm
- 420×300×540 mm
- 系统重量:
- 27,7 kg
- 25,7 kg