Methanol
2865, Glass bottle, 1.000 mL
目录编号 HACHL322.1000
供应商: HACH
Jump to:
Product Details & Documents
同义词: 木精
- CAS号:67-56-1
- 化学式:CH₃OH
- MDL 编号:MFCD00004595
- Molecular Weight:32.04 g/mol
- 默克索引:13,05984
- 沸点:64,6 °C (1013 hPa)
- 熔点:–98 °C
- 密度:0,7918 g/cm³ (20 °C)
- 闪点:11 °C (closed cup)
- 储存温度:Ambient
- EINECS:200-659-6
- UN:1230
- ADR:3,II
- REACH:01-2119433307-44
规格
规格
- 包装类型:Glass bottle
- 包装规格:1.000 mL
- 顺序:2865
- Cat. No.:HACHL322.1000
- 环境认证:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health
规格
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



