Orders LinkContactUsLinkComponent
Buffer, TRIS solution pH 7,5 (1 mol/L)
Buffer, TRIS solution pH 7,5 (1 mol/L)
TRIS buffer solution pH 7,5 (1 mol/l), Glass bottle, 1 L
目录编号 INVT15567027
Marketsource
供应商:   INVITROGEN
Buffer, TRIS solution pH 7,5 (1 mol/L)
目录编号 INVT15567027
供应商:  INVITROGEN
Marketsource
CAS号:  
Buffer, TRIS solution pH 7,5 (1 mol/L)
TRIS buffer solution pH 7,5 (1 mol/l), Glass bottle, 1 L
Jump to:
CAS号:  
Price
询价
Log in 查看库存
1 * 1 升
Product Details & Documents
规格
规格
  • 包装类型:
    Glass bottle
  • 包装规格:
    1 L
  • 说明:
    TRIS buffer solution pH 7,5 (1 mol/l)
规格