载玻片盒,Q Path®
载玻片盒, 半透明材质, 塑料, 5, 82×30×18 mm
目录编号 VWRI631-1079
供应商: VWR Collection
Jump to:
Product Details & Documents
This box for five slides prevents items from breaking or sticking. Can be used for applications in cytology, such as conventional smears.
W×D×H: 82×30×18 mm
规格
- 滑道数量:5
- 颜色:半透明材质
- 说明:载玻片盒
- 材质:塑料
- 宽度×深度×高度:82×30×18 mm