载玻片柜,Q Path®
载玻片柜,容纳1000个载玻片, 自然色, 卡纸板, 1000, 84×155×310 mm
目录编号 VWRI631-1066
供应商: VWR Collection
Jump to:
Product Details & Documents
This cabinet, with lid, can be used in cytology and histology applications.
- Simple design for easy handling
- Quick handling of microscope slides
- Able to hold a large number of items
W×D×H: 84×155×310 mm
Documents
规格
- 滑道数量:1000
- 颜色:自然色
- 说明:载玻片柜,容纳1000个载玻片
- 材质:卡纸板
- 宽度×深度×高度:84×155×310 mm