规格
- 容量:1 ml
- 说明:Protein G GraviTrap™
- 顺序:01
- 结合量:Approx. 20 mg human IgG/column
- 硬件栏:PP
- 包装规格:10
规格
关于此项目
Protein G GraviTrap includes ten columns with 1 ml of resin for general purpose purification of polyclonal and monoclonal antibodies from most species.
- Broad binding specificity – binds a broad range of IgG species and subclasses from most species including rat
- High binding capacity – approximately 20 mg human IgG/ml resin
- Simple – GraviTrap prepacked gravity flow columns enable simple one-step purification of proteins in 30 min without the need for equipment
Protein G GraviTrap columns are prepacked with Protein G Sepharose 4 Fast Flow resin. These gravity flow columns are designed for fast and efficient manual purification of monoclonal and polyclonal antibodies. Each of these protein G columns is reusable up to five times. Prepacked gravity flow columns provide convenient, fast, and reliable results.