HPLC columns, Acclaim™ 120 C8
目录编号 HYPE059141
规格
- 列格式:Analytical
- 内径:4,60 mm
- 颗粒大小:5 µm
- 孔大小:120 Å
- 长度:250 mm
- 加端盖的:是
- 美国药典代码:L7
- pH 范围:2 - 8
- 表面积:300 m²/g
- 温度限值:60 °C
- 碳负载:11%
- Phase:C8
- Packing material:Spherical, fully porous ultrapure silica
- 包装规格:1
规格
关于此项目
The Acclaim™ 120 C8 high performance reversed-phase columns are ideal for the separation of small molecules and this can be used for a variety of applications in the pharmaceutical, environmental, food and many other industrial sectors.
- Low silanol activity for excellent peak shapes for basic analytes
- Reproducible column-to-column performance
- Extremely Low bleed, fully compatible with MS