
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride)
Catalog # CHLCR01315060111
Supplier: LOCAL DEALS
CAS Number:
Specifications
- 包装类型:Glass bottle for solids
- 包装规格:1 kg
- 顺序:4024
Specifications
About this item
:
:191.7 g/mol
Melting Pt:112…113 °C
EINECS:247-361-2
:GHS07
Storage Temperature:Freezer
CAS Number:25952-53-8
Formula:C₈H₁₇N₃·HCl
MDL Number:MFCD00012503