Order Entry
China
ContactUsLinkComponent
4-异丙氧基苯硼酸 (contains varying amounts of Anhydride)
Irritant
4-异丙氧基苯硼酸 (contains varying amounts of Anhydride)
Catalog # TCIAI0806-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
4-异丙氧基苯硼酸 (contains varying amounts of Anhydride)
Catalog # TCIAI0806-5G
Supplier:  TCI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • 包装类型:
    Glass bottle
  • 包装规格:
    5 g
  • 顺序:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (Neutralization titration):
    97.0 to 112.0 %

Specification Test Results

About this item

 :
 :180.01 g/mol
 :GHS07
CAS Number:153624-46-5
Formula:C₉H₁₃BO₃
MDL Number:MFCD03427051