Order Entry
China
Orders LinkContactUsLinkComponent
Banner_28702.jpg
Enzymatic Methyl-seq (EM-seq™): A new option for 5mC and 5hmC identification
Webinar
Thursday, 29th October, 2020 
14.00 - 15.00 EST
Replay
 Enzymatic Methyl-seq (EM-seq™): A new option for 5mC and 5hmC identification | Avantor
Email_Icon_RGB.png
Email alerts
Registration_Icon_RGB.png
Free registration
Chat_Icon_RGB.png
Chat area
Certified_Icon_RGB.png
Certificate of attendance
Increase accuracy and sensitivity of methylation detection
Identification of 5mC and 5hmC in DNA increases insight into gene regulatory mechanisms. Bisulfite sequencing has been commonly used for this, but the chemical-based conversion damages and degrades DNA, introducing bias into the data. In this webinar we will discuss NEBNext® Enzymatic Methyl-seq (EM-seq™), an enzyme-based method that minimizes damage to DNA. This method enables longer insert sizes, lower duplication rates and reduced GC bias, resulting in more accurate and sensitive methylation detection.
Presented by
Siva-Chavadi.png
Siva Chavadi
Siva Chavadi received his Ph.D. from the University of Hyderabad, India and his graduate and postdoctoral work utilized functional genomics-based tools to develop diagnostics, anti-bacterials and vaccines against TB.  Siva is now a Senior Field Application Scientist at New England Biolabs, focusing on next generation sequencing.