0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
384 well 140uL Polypropylene, Long drip Glass fibre, mean pore size 0.7µm 1 * 10 items
Catalog # VWRI732-4430
Supplier:
VWR Collection
New Product
Expand image
384 well 140uL Polypropylene, Long drip Glass fibre, mean pore size 0.7µm 1 * 10 items
Catalog # VWRI732-4430
Supplier:
VWR Collection
New Product