0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Grinding jar EasyFit for Planetary Ball Mills, tungsten carbide, 250ml 1 * 1 items
Catalog # RETS01.462.0497
Supplier:
RETSCH
Expand image
Grinding jar EasyFit for Planetary Ball Mills, tungsten carbide, 250ml 1 * 1 items
Catalog # RETS01.462.0497
Supplier:
RETSCH