Pyrethrins (technical)
Pyrethrins (technical)
EHERCA16620000
- SMILES: C/C(C)=C/C1C(C)(C)[C@@H]1C(O[C@@H]2C(C)=C(C/C=C/C=C)C(C2)=O)=O.C/C(C([CH3]=O)=O)=C\C3C(C)(C)[C@@H]3C(OC4C(C)=C(C/C=C/C=C)C(C4)=O)=O
- Packing group: III
- Description:Pyrethrins (technical)
- UN No.:3082
- Cat. No.:EHERCA16620000
- Storage conditions:–18 °C
- Hazard class description:GHS07, GHS09
- GHS signal:Warning
- Synonym(s):Pyrethrins;Pyrethrins and Pyrethroids;Biopiren plus;Chrysanthemates pyrethrins and pyrethroids;Diatect;Firmotox;Goldgeist forte;Nettafid;Parexan;Piresan;Pyganic;Pyganic MUP 20;Pyre-sin Plus;Pyrellin;Pyrethrine;Pyrethrines;Pyrethrins and Pyrethroids, chrysanthemates;Pyrethrins and Pyrethroids, esters;Pyrethrins, chrysanthemates;Pyrethro Vioryl 5SC;Pyrethrum;Pyrethrum (insecticide);Pyrethrum FS;Pyrevert;Pyrocide 50;Spruzit;Take Down;Take Down Garden Spray
- CAS No.:8003-34-7
- Shipping conditions:Ice pack (–20 °C)



