Order Entry
China
ContactUsLinkComponent
Furnace MuFFle lt5/14/b510 tMax1400 with with liFt Door Door anD Protective gas connection anD over-teMPerature liMiter 1 * 1 items
Catalog # NABEL-055H4ANLGT
Supplier:  NABERTHERM
Furnace MuFFle lt5/14/b510 tMax1400 with with liFt Door Door anD Protective gas connection anD over-teMPerature liMiter 1 * 1 items
Catalog # NABEL-055H4ANLGT
Supplier:  NABERTHERM