Bottle brushes
White, Polyester, 101,6 mm, Plastic, 762 mm
Catalog # MICRBRWH-70
Supplier: MICRONOVA
Jump to:
Product Details & Documents
Brushes with polyester bristles and plastic handle for cleaning bottles and other glassware.
- Base resins used to produce blocks and bristles are made from FDA approved materials
- BPA-free
Specifications
- Bristle material:Polyester
- Brush head Ø:101,6 mm
- Handle material:Plastic
- Description:White
- Total length:762 mm



