Order Entry
China
ContactUsLinkComponent
SwitchBox: control unit for SCAT Built-in Signalbox or Signalbox T1/T5. Controls electrically operated valve depending on the full-s tate signal. Aditionally, SwitchBox can be used in any kind of automation circuit (pump, valve, etc. control). 1 * 1 items
  SCTV106730
undefined
SwitchBox: control unit for SCAT Built-in Signalbox or Signalbox T1/T5. Controls electrically operated valve depending on the full-s tate signal. Aditionally, SwitchBox can be used in any kind of automation circuit (pump, valve, etc. control). 1 * 1 items
  SCTV106730