Order Entry
China
ContactUsLinkComponent
Biotinylated Human C1q R1, His,Avitag (C11-H82E9) is expressed from human 293 cells (HEK293). It contains AA Thr 22 - Lys 580 (Acces sion # Q9NPY3-1). 1 * 200 µG
Catalog # ACRBC11-H82E9200UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Human C1q R1, His,Avitag (C11-H82E9) is expressed from human 293 cells (HEK293). It contains AA Thr 22 - Lys 580 (Acces sion # Q9NPY3-1). 1 * 200 µG
Catalog # ACRBC11-H82E9200UG
Supplier:  ACROBIOSYSTEMS