Order Entry
China
ContactUsLinkComponent
Biotinylated Human DDR1, His,Avitag™ (DD1-H82E9) is expressed from human 293 cells (HEK293). It contains AA Asp 21 - Thr 416 (Access ion # Q08345-1). 1 * 25 µG
Catalog # ACRBDD1-H82E9-25UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Human DDR1, His,Avitag™ (DD1-H82E9) is expressed from human 293 cells (HEK293). It contains AA Asp 21 - Thr 416 (Access ion # Q08345-1). 1 * 25 µG
Catalog # ACRBDD1-H82E9-25UG
Supplier:  ACROBIOSYSTEMS