Order Entry
China
ContactUsLinkComponent
Masterflex® Fitting, PVDF, Elbow, Hose Barb Union, 1/2" ID"; 10/PK
Masterflex® Fitting, PVDF, Elbow, Hose Barb Union, 1/2" ID"; 10/PK
Catalog # MFLA40614-86
undefined
Masterflex® Fitting, PVDF, Elbow, Hose Barb Union, 1/2" ID"; 10/PK
Catalog # MFLA40614-86
  • Orifice dimensions:
  • 8,73 mm (0,34375")
  • 5,56 mm (0,21875")
  • 11,9 mm (0,46875")
  • 0,09375"
  • 0,0625"
  • 3,175 mm (0,125")
  • 0,09375"
  • 19,84 mm (0,78125")
  • 14,29 mm (0,5625")
  • Sterilisation:
  • Autoclave or ETO
  • Autoclave or ETO
  • Autoclave or ETO
  • Autoclave or ETO
  • Autoclave or ETO
  • Autoclave or ETO
  • Autoclave or ETO
  • Autoclave or ETO
  • Autoclave or ETO
  • Autoclave or ETO
  • Currently Viewing
Specifications
About this item
Quick and easy assembly without tools.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Create 90° bends in flexible tubing—without the kinks!
  • Barb size/tubing int.Ø (port 1):
    1/2"
  • Barb size/tubing int.Ø (port 2):
    1/2"
  • Connection Type:
    Barbed Union
  • Flow pattern:
    Elbow
  • Description:
    Fitting, PVDF, Elbow, Hose Barb Union, 1/2" ID"; 10/PK
  • Environmentally Preferable:
  • Orifice dimensions:
    8,73 mm (0,34375")
  • Sterilisation:
    Autoclave or ETO
  • Pk:
    10
  • Fitting material:
    PVDF