Order Entry
China
ContactUsLinkComponent
Masterflex® L/S® Easy-Load® Pump Head with Long-Shaft for Precision Tubing, 6-Roller, PARA Housing, SS Rotor
Masterflex® L/S® Easy-Load® Pump Head with Long-Shaft for Precision Tubing, 6-Roller, PARA Housing, SS Rotor
Catalog # MFLA07514-60
2 year warranty
undefined
Masterflex® L/S® Easy-Load® Pump Head with Long-Shaft for Precision Tubing, 6-Roller, PARA Housing, SS Rotor
Catalog # MFLA07514-60
2 year warranty
About this item
Save time with fast tubing changes and reduced maintenance.
  • New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
  • Rotor is constructed of stainless steel (SS) for enhanced durability
This L/S® Easy-Load® II Pump Head, when combined with a Masterflex® L/S® drive or compatible system, is designed to provide a simple, easy-to-use peristaltic pump system. The pump head accepts several different tubing sizes for a wide range of flow rates.
  • No. of channels:
    1
  • Pump series:
    L/S
  • Description:
    L/S® Easy-Load® Pump Head with Long-Shaft for Precision Tubing, 6-Roller, PARA Housing, SS Rotor
  • Seq:
    03
  • Environmentally Preferable:
  • Model:
    07514-60
  • Occlusion:
    Fixed
  • Open-head sensor:
    No
  • Tubing retention:
    Automatic
  • Tubing sizes accepted:
    L/S® 13, L/S 14, L/S 16, L/S 25
  • Warranty:
    2 years
  • Pk:
    1
  • Rollers:
    6
  • Housing material:
    Polyarylamide (PARA)
  • Rotor material:
    Stainless Steel