Order Entry
China
ContactUsLinkComponent
Smart Gasket G-TH-075-E-2 3/4" Tri-Clamp Gasket, 2 Port, EPDM; 1/Ea
Smart Gasket G-TH-075-E-2 3/4" Tri-Clamp Gasket, 2 Port, EPDM; 1/Ea
Catalog # MFLB30543-05
Supplier:  MASTERFLEX PREMIUM
undefined
Smart Gasket G-TH-075-E-2 3/4" Tri-Clamp Gasket, 2 Port, EPDM; 1/Ea
Catalog # MFLB30543-05
Supplier:  MASTERFLEX PREMIUM
About this item
Smart Gasket lets you take samples or readings from a sanitary line.
  • Measure temperature, take a sample, or dose chemicals into your process line
  • Install in between two sanitary fittings
The Smart Gasket is used as a standard sanitary gaskets with standard Tri-clamp® fittings. The gaskets have molded ports that allow either a thermocouple or sampling tubes to be inserted into the line. Order the appropriate Smart Gasket clamp separately.
  • Sanitary clamp size (Port 1):
    3/4"
  • Connection type:
    Sanitary
  • Description:
    3/4" Tri-Clamp, 2 Port, EPDM, Each
  • Seq:
    17
  • Environmentally Preferable:
  • Pk:
    1
  • Fitting material:
    EPDM
  • Sanitary gasket size:
    3/4"
  • No. of ports:
    2
  • Warranty:
    1 year