Order Entry
China
ContactUsLinkComponent
Masterflex® L/S® Variable-Speed Digital Drive System with PTFE-Diaphragm Head; 90 to 260 VAC
Masterflex® L/S® Variable-Speed Digital Drive System with PTFE-Diaphragm Head; 90 to 260 VAC
Catalog # MFLA77915-10-EU
CE Compliant
ISO9001
undefined
Masterflex® L/S® Variable-Speed Digital Drive System with PTFE-Diaphragm Head; 90 to 260 VAC
Catalog # MFLA77915-10-EU
CE Compliant
ISO9001

关于此项目

Ideal for chemical feed/metering and applications requiring high-purity, high-pressure or both.

  • Flow range 80 to 800 ml/min—flow rate depends on drive rpm and tubing size
  • PTFE wetted parts help maintain fluid purity
  • Pump through filters or into pressurized system
  • Check valve on inlet side of pump holds prime for up to two hours when pump is off
  • Unique geometry minimizes fluid churning and dead volume for high metering accuracy without reagent contamination
  • Accurate and repeatable settings with 3-digit LED display of rpm
  • Maximize processes in a minimal footprint by stacking splash-resistant, IP33-rated drives
  • Two ways to control speed, direction, and start/stop—use either the membrane keypad or remote-control capabilities

These L/S® precision variable-speed console drives offer precise ±0.25% speed control for accurate flow regulation. The large LED display and LED indicators confirm motor rpm and motor direction at a glance. The complete pump systems include all you need to start pumping.

Delivery: Includes pTFE-diaphragm pump head 07090-42, 12 ft (3.6-m) of PTFE tubing (1/4" ID x 3/8" OD), L/S drive 07528-10, two PFA pipe adapters (1/4" NPT(F) x 3/8" OD), foot valve, mounting tools, and coupling hardware.
  • Maximum Flow Rate:
    800 ml/min
  • Power:
    90 - 260 VAC; 50/60 Hz; 1,8 A @ 115 V; 1,2 A @ 230 V
  • Accuracy:
    ±0,25% (1 rpm at 600 and 300 rpm; 0,1 rpm at 100 rpm)
  • Control type:
    Variable-Speed Digital
  • Min. flow rate:
    80 ml/min
  • No. of channels:
    1
  • Pump series:
    L/S
  • Description:
    L/S PTFE-Diaphragm Pump System; 6 to 600 rpm, 90 to 260 VAC
  • Height:
    21,59 cm
  • Length:
    43,94 cm
  • Environmentally Preferable:
  • Duty cycle:
    Continuous
  • Drive IP rating:
    IP 33
  • Min. RPM:
    6 rpm
  • Open-head sensor:
    No
  • Remote I/O:
    4 to 20 mA, 0 to 10 V, start/stop and direction via contact closure
  • Tubing sizes accepted:
    L/S® 13, 14, 16, 25, 17, 18
  • Washdown capable:
    No
  • Tubing:
    06605-15, 12-ft (PTFE Tubing, 1/4" x 3/8")
  • Drive:
    07528-10 (L/S® Variable-Speed Digital Drive with Remote I/O, 600 rpm)
  • Warranty:
    2 years
  • Pk:
    1
  • Pump head:
    07090-42 (L/S® PTFE-Diaphragm Pump Head)
  • Width:
    21,08 cm
  • Housing material:
    ABS plastic and aluminium
  • Electrical plug type:
    EU
  • Max. RPM:
    600 rpm