Micropump Drive for A-Mount pump heads, explosion-proof (XPRF), 3150 rpm, 115/230 VAC
Catalog # MFLB07144-95
Supplier: MASTERFLEX PREMIUM
About this item
A variety of low-cost drives for numerous applications.
- Magnetic coupling minimizes friction—all horsepower is transmitted directly to the gears
- Hundreds of hours of dependable leakproof service
These Economical Drives for Micropump A-Mount Pump Heads are designed with magnetic coupling to minimize friction. Choose from AC motor drives, DC motor drives, explosion-proof drives, or air-powered drives.
Explosion-proof and air-powered models are ideal for safe pumping in hazardous areas. Explosion-proof drives are rated for NEMA Class 1, Groups C and D locations.
- Power:115/230 VAC; 1,8 A
- Description:Drive for A-Mount pump heads, explosion-proof (XPRF), 3150 rpm, 115/230 VAC
- Height:133 mm
- Length:27,9 cm
- Environmentally Preferable:
- Duty cycle continuous:Yes
- Motor enclosure type:PSC
- Motor hp:0,17
- Min. temperature:4 °C (40 °F)
- Max. temperature:60 °C (140 °F)
- Warranty:1 year
- Pk:1
- Width:15,2 cm
- Max. motor RPM:3150 rpm