Masterflex® I/P® Easy-Load® Main Body Assembly, Occlusion Bed and Rear Bearings, PPS Housing, SS Rotor
Catalog # MFLA77601-67
Supplier: MASTERFLEX PROPRIETARY
About this item
Get the most our of your Masterflex® I/P® Easy-Load® Pump Head with these replacement parts.
- Suitable for use with pump heads 77601-60 and 77601-62
Keep your pump head operating at its best with these replacement main body and occusion bed assemblies.
- Pump series:I/P
- Description:Main body/occlusion bed assemblies with rear bearing, PPS/SS housing material.
- Environmentally Preferable:
- Pk:1
- Housing material:PPS
- Rotor material:Polyphenylene Sulfide (PPS)