Masterflex® L/S® Easy-Load® II Pump Head Replacement Rotor Assembly for High-Performance Precision Tubing, CRS
Catalog # MFLA77200-59
Supplier: MASTERFLEX PROPRIETARY
About this item
Get the most our of your Masterflex® L/S® Easy-Load® II Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® pump head
Replace your pump head rotor assembly to extend the life of your pump head.
- Pump series:L/S
- Description:Replacement rotor assembly for L/S Easy-Load II pump heads 77200-52,
- Environmentally Preferable:
- Warranty:2 years
- Pk:1
- Rotor material:Cold-rolled steel