Masterflex® 4 Multi Rack for Syringe Pump, Infusion and Withdrawal Design
Catalog # MFLA74905-61
Supplier: MASTERFLEX PROPRIETARY
About this item
Make the most of your Masterflex® syringe pumps with these accessories.
- Optimize performance of your Masterflex pumps
Parts and accessories for Masterflex syringe pumps.
- Description:4 Multi Rack for Syringe Pump, Infusion and Withdrawal design
- Environmentally Preferable:
- Model:74905-61
- Syringe size accepted:60 - 140 ml
- Warranty:1 year
- Pk:1
- No. of syringes:4