Masterflex® Touch Screen Syringe Pump Continuous Flow Starter Kit
Catalog # MFLA74901-90
Supplier: MASTERFLEX PROPRIETARY
About this item
Make the most of your syringe pump with these accessories.
- Choose from a wide range of replacement parts and accessories
Optimize performance of your Masterflex® Touch-Screen, Continuous-Cycle, Four-Syringe Pump
Includes 2 check valves, two 10 mL plastic syringes, and a short length of tubing.
- Description:Syringe Pump Accessories, Continuous Flow Starter Kit
- Environmentally Preferable:
- Warranty:1 year
- Pk:1