Order Entry
China
ContactUsLinkComponent
Masterflex® Fitting, Polyethylene, Straight, Thread Reducer, 3/8" NPT(M) x 1/2" NPT(M); 10/PK
  MFLA30623-52
CE certificate
undefined
Masterflex® Fitting, Polyethylene, Straight, Thread Reducer, 3/8" NPT(M) x 1/2" NPT(M); 10/PK
  MFLA30623-52
CE certificate
  • Model:
  • 30623-52
  • 30623-40
  • 30623-50
  • 30623-36
  • 40623-39
  • 40623-47
  • 30627-54
  • 30623-46
  • 30623-48
  • 30623-42
  • 30623-54
  • 30623-41
  • 40623-35
  • 40623-55
  • 40623-43
  • 40623-51
  • 40627-43
  • 30623-38
  • 40627-55
  • 40627-35
  • 31701-89
  • 30623-34
  • 31701-88
  • 30623-32
  • Sterilisation:
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Ethylene oxide (ETO) only
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) only
  • Warranty:
  • 1 year
  • 1 year
  •  
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 3/8" NPT(M) x 1/2" NPT(M)
  • Constructed of durable polyethylene
Threaded fittings provide a more permanent fluid path. Use these fittings to reduce or enlarge connection sizes. Choose the size and material to meet your application needs.
  • Connection Type:
    Threaded Reducer
  • Flow pattern:
    Straight
  • Thread size NPT(M) (port 1):
    ⅜"
  • Thread size NPT(M) (port 2):
    ½"
  • Description:
    Fitting, Polyethylene, Straight, Thread Reducer, ⅜" NPT(M) x ½" NPT(M); 10/PK
  • Seq:
    17
  • Environmentally Preferable:
  • Model:
    30623-52
  • Sterilisation:
    Ethylene oxide (ETO) only
  • Pk:
    10
  • Fitting material:
    HDPE