Order Entry
China
ContactUsLinkComponent
Barbed Fittings; Reducing Connector, Stainless Steel, 1/2" x 1/4" Tubing ID
Barbed Fittings; Reducing Connector, Stainless Steel, 1/2" x 1/4" Tubing ID
Catalog # MFLA30620-11
undefined
Barbed Fittings; Reducing Connector, Stainless Steel, 1/2" x 1/4" Tubing ID
Catalog # MFLA30620-11
  • Model:
  • 30620-11
  • 30620-12
  • 30620-09
  • 31208-14
  • 30620-10
  • 31208-15
  • 31208-13
  • Orifice dimensions:
  • 4,763 mm (0,1875")
  • 8,73 mm (0,34375")
  • 0,7925 mm (0,03125")
  • 4,763 mm (0,1875")
  • 0,7925 mm (0,03125")
  • Sterilisation:
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Warranty:
  • 1 Month
  • 1 Month
  • 1 year
  • 1 Month
  • 1 year
  • 1 year
  • Currently Viewing
Specifications
About this item
Ideal for low-pressure applications.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi). Just slip your flexible tubing or hose onto the barb to create a secure seal. 316 stainless steel material features very good chemical resistance with a wide temperature range. Sterilise via ethylene oxide, autoclave, or gamma irradiation.
  • Barb size/tubing int.Ø (port 1):
    ½"
  • Barb size/tubing int.Ø (port 2):
    ¼"
  • Connection Type:
    Barbed reducer
  • Flow pattern:
    Straight
  • Description:
    Barbed fittings, reducing connector, stainless steel, tubing ID: ½×¼"
  • Seq:
    01
  • Environmentally Preferable:
  • Model:
    30620-11
  • Orifice dimensions:
    4,763 mm (0,1875")
  • Sterilisation:
    Autoclave, ethylene oxide (ETO), or gamma
  • Warranty:
  • Pk:
    1
  • Fitting material:
    Stainless steel 316