Masterflex® Fitting, Polycarbonate, Four Ports, Manifolds with Female Luer Locks, 180° Rotation, Non-Sterile; 10/PK
Catalog # MFLA30600-42
Supplier: MASTERFLEX PROPRIETARY
About this item
Regulate flow without any leaking.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Polycarbonate manifolds have polyurethane stems and HDPE handles. Ports have female luer locks.
- Flow pattern:4 Ports, 180 Degree Rotation
- Description:Fitting, Polycarbonate, Four Ports, Manifolds with Female Luer Locks, 180° Rotation, Non-Sterile; 10/PK
- Environmentally Preferable:
- Wetted materials:Polycarbonate
- Warranty:Out of box
- Pk:10
- Inlet port:Female Luer
- Outlet port:Female Luer