Order Entry
China
ContactUsLinkComponent
SaniSure SS-CUT-170 Single-Use Sanitary Clamp Cable Tie Removal Tool
SaniSure SS-CUT-170 Single-Use Sanitary Clamp Cable Tie Removal Tool
Catalog # MFLA30562-42
undefined
SaniSure SS-CUT-170 Single-Use Sanitary Clamp Cable Tie Removal Tool
Catalog # MFLA30562-42
About this item
Fast, safe, secure, and cost-effective connections!
  • Even 360-degree compression to prevent leakage
  • Withstands pressures up to 100 psi (6.9 bar)
  • Reduces installation time
  • Ergonomic design and padded grips
  • Fit blade under the SaniSure® CLAMP securing cable tie and squeeze for easy zip tie removal
SaniSure clamps feature an innovative design for fast, safe, secure connections and offer a cost-effective solution for connecting single-use installations. Made of USP Class VI-approved nylon, these clamps are designed for single use applications but are durable enough to withstand repetitive use. Unlike traditional two-point load stainless steel sanitary clamps, the design of these clamps ensure even 360-degree compression over the entire fitting surface—no more leaks and no gasket intrusion in your process line! Their low-profile rounded edges reduce the risk of penetrating bag assemblies, and the evenly controlled compression allows easy connection for plastic-to plastic, plastic-to-stainless steel, or stainless steel-to-stainless steel coupling joints.
To use, simply insert the cable tie end in its mate a couple clicks; position clamp on ferrule; hold both clamp parts together; move clamp around the tri-clamp fitting joint and slide clamp closed. Complete the closure by using the clamping tool to bring it to the proper tension. The easy-to-use design reduces chances of risk associated with carpal tunnel or other wrist injuries and yields a tamper-proof connection that is not operator dependent. As a single-use device, they eliminate need for cleaning validations and reduce your overall downtime for installation.
  • Description:
    Sanitary Clamp/Cable Tie Removal Tool
  • Environmentally Preferable:
  • Compatible with:
    SaniSure® CLAMP sanitary fitting clamps
  • Pk:
    1
  • Clamp type:
    Accessories
  • Warranty:
    3 Months