Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Recombinant Anti-DDR1 Antibody, Rabbit Monoclonal 1 * 100 µl
Catalog # SINO10730-R203
Supplier:
Sino Biological
Expand image
Recombinant Anti-DDR1 Antibody, Rabbit Monoclonal 1 * 100 µl
Catalog # SINO10730-R203
Supplier:
Sino Biological