Order Entry
China
ContactUsLinkComponent
Steel frame slides with PET membrane (1.4 µm), 50 pieces, 26x76 mm area for LMD applications null
Catalog # LEIM11505151
Supplier:  LEICA MICROSYSTEMS
Steel frame slides with PET membrane (1.4 µm), 50 pieces, 26x76 mm area for LMD applications null
Catalog # LEIM11505151
Supplier:  LEICA MICROSYSTEMS