0
¥0.00
Order Entry
登录
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
一次性塑料滴管 5ML 205MM 分度值:1ML 100支/包 1 * 100 件
Catalog # CHLCQR7329
Supplier:
LOCAL DEALS
展开图片
一次性塑料滴管 5ML 205MM 分度值:1ML 100支/包 1 * 100 件
Catalog # CHLCQR7329
Supplier:
LOCAL DEALS