Order Entry
China
Orders LinkContactUsLinkComponent
Volumetric flasks, class B
25 ml, 10/19, 0,08 ± ml
Catalog # VITL671941
Supplier:   VITLAB
Volumetric flasks, class B
Catalog # VITL671941
Supplier:  VITLAB
Volumetric flasks, class B
Volumetric flasks, class B
25 ml, 10/19, 0,08 ± ml
Price
Inquire For Price
Log in for availability
1 * 2 件
Product Details & Documents
PP, with PP stoppers.
Highly transparent in accordance with DIN 12664 (glass) with blue ring mark and lettering. Individually adjusted mark to contain (TC, In).
Specifications
  • NS:
    10/19
  • Capacity:
    25 ml
  • Seq:
    02
  • Tolerance:
    0,08 ± ml