0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
枯草芽孢杆菌 CMCC(B) 63501 ,Microball 杭州微球 定量菌种 浓度100-1000cfu /0.1ml 1 * 0,1 毫升
Catalog # CHLCMGP-D1-C3
Supplier:
LOCAL DEALS
Expand image
枯草芽孢杆菌 CMCC(B) 63501 ,Microball 杭州微球 定量菌种 浓度100-1000cfu /0.1ml 1 * 0,1 毫升
Catalog # CHLCMGP-D1-C3
Supplier:
LOCAL DEALS