Order Entry
China
Orders LinkContactUsLinkComponent
Thread adaptors, TYPE B, b.safe
Thread adaptors, TYPE B, b.safe
PP, GL 45 (male), G 2"/BSP 2" (male)
Catalog # BOHLM615-01
Supplier:   BOHLENDER
Thread adaptors, TYPE B, b.safe
Catalog # BOHLM615-01
Supplier:  BOHLENDER
Thread adaptors, TYPE B, b.safe
Thread adaptors, TYPE B, b.safe
PP, GL 45 (male), G 2"/BSP 2" (male)
Price
Inquire For Price
Log in for availability
1 * 1 件
Product Details & Documents
All adaptors of this type are supplied with two male threads and recessed grips.
  • Stay flexible and independent
  • With recessed grips for an easy assembly
Your existing caps and waste caps do not fit with the flasks and canisters which you would like to use or vice versa? Simply use b.safe thread adaptors for a flexible combination of caps and waste caps independently from the thread on the flask and/or container.
Specifications
  • Thread:
    GL 45 (male), G 2"/BSP 2" (male)
  • Material:
    PP
  • Seq:
    01