Order Entry
China
ContactUsLinkComponent
Glass slides with PEN membrane(4µm), 50 pieces, 26x76xmm area for LMD7000 applications temperature resistant up to 70°C limited suit ability for microwave treatment water (up to 5 min), xylene (up to 15min),methanol,ethanol,acetone (5 min) resistant 1 * 1 items
Catalog # LEIM11600288
Supplier:  LEICA MICROSYSTEMS
Glass slides with PEN membrane(4µm), 50 pieces, 26x76xmm area for LMD7000 applications temperature resistant up to 70°C limited suit ability for microwave treatment water (up to 5 min), xylene (up to 15min),methanol,ethanol,acetone (5 min) resistant 1 * 1 items
Catalog # LEIM11600288
Supplier:  LEICA MICROSYSTEMS