0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
碱性液体清洗剂,浓缩型,无泡沫,不含磷、氯、NTA及EDTA。 1 * 4 件
Catalog # CHLCRBSA156
Supplier:
LOCAL DEALS
Expand image
碱性液体清洗剂,浓缩型,无泡沫,不含磷、氯、NTA及EDTA。 1 * 4 件
Catalog # CHLCRBSA156
Supplier:
LOCAL DEALS