LABGENE 15/50 ml Magnetic Separation Combo Rack, designed for large separations of common magnetic bead chemistries and protocols wi th up to 3 x 50 ml tubes and or 3 x 15 ml tubes 1 * 1 ST
Catalog # MSPPMSR1550
Supplier: MARKET SOURCE PART PROCESS