Order Entry
China
ContactUsLinkComponent
温度变送器TL21{50 TO 150}C Brurns Transmitter for use with = Three Wire Single, RTD Transmitter Model = TL21: 2 Wire Analog (0.00385 Coefficient) Transmitter Range = {50 to 150} °C Availability = 1 Week ARO 1 * 1 件
Catalog # IBSATL21-M50TO150C
Supplier:  IBS STOCK ITEMS
温度变送器TL21{50 TO 150}C Brurns Transmitter for use with = Three Wire Single, RTD Transmitter Model = TL21: 2 Wire Analog (0.00385 Coefficient) Transmitter Range = {50 to 150} °C Availability = 1 Week ARO 1 * 1 件
Catalog # IBSATL21-M50TO150C
Supplier:  IBS STOCK ITEMS