VWR®, Sampling Spoons
170 mm, 10 ml
VWRU10819-505
PS, white. These disposable spoons are produced in a cleanroom environment and sterilised by gamma radiation. They are ideal for sampling powder, granules and pastes.
- Available in two different sizes: 2,5 ml (similar to a teaspoon) and 10 ml (similar to a tablespoon)
- Production, assembly and packaging according to cleanroom Class 7 (10000)
- According to EU food and FDA guidelines
- Individually packed, sterilised by gamma radiation
- Capacity:10 ml
- Length:170 mm
- Seq:03



