Order Entry
China
ContactUsLinkComponent
ActiveMax® Mouse VEGF164, Tag Free (HPLC-verified) (VE4-M4216) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - A rg 190 (Accession AAB22253.1 ). 1 * 20 µG
  ACRBVE4-M4216-20UG
ActiveMax® Mouse VEGF164, Tag Free (HPLC-verified) (VE4-M4216) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - A rg 190 (Accession AAB22253.1 ). 1 * 20 µG
  ACRBVE4-M4216-20UG