Order Entry
China
ContactUsLinkComponent
Biotinylated Human CD4, His,Avitag (CD4-H82E8) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Pro 396 (Accessio n P01730-1). 1 * 25 µG
Catalog # ACRBCD4-H82E8-25UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Human CD4, His,Avitag (CD4-H82E8) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Pro 396 (Accessio n P01730-1). 1 * 25 µG
Catalog # ACRBCD4-H82E8-25UG
Supplier:  ACROBIOSYSTEMS