Order Entry
China
ContactUsLinkComponent
Human IL-1RL1, His Tag (IL1-H5229) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - Phe 328 (Accession Q01638-2). 1 * 200 µG
Catalog # ACRBIL1-H5229200UG
Supplier:  ACROBIOSYSTEMS
Human IL-1RL1, His Tag (IL1-H5229) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - Phe 328 (Accession Q01638-2). 1 * 200 µG
Catalog # ACRBIL1-H5229200UG
Supplier:  ACROBIOSYSTEMS