Order Entry
China
ContactUsLinkComponent
Side access rack system to complete the SU780UE/XLE upright freezer 2inches boxes with 100 place dividers (30 racks, 600 box capaci ty) BOXES and DIVIDERS INCLUDED 1 * 1 items
Catalog # GLCL780-RS6-RACK
Supplier:  GLOBAL COOLING
Side access rack system to complete the SU780UE/XLE upright freezer 2inches boxes with 100 place dividers (30 racks, 600 box capaci ty) BOXES and DIVIDERS INCLUDED 1 * 1 items
Catalog # GLCL780-RS6-RACK
Supplier:  GLOBAL COOLING