Order Entry
China
ContactUsLinkComponent
Kern EWJ 3000-2 precision balance, weighing capacity 3000, accuracy: 0.01 g, precision: ±0.05 g 1 * 1 items
Catalog # KERNEWJ3000-2
Supplier:  KERN SOHN
Kern EWJ 3000-2 precision balance, weighing capacity 3000, accuracy: 0.01 g, precision: ±0.05 g 1 * 1 items
Catalog # KERNEWJ3000-2
Supplier:  KERN SOHN