Order Entry
China
ContactUsLinkComponent
Implen NanoPhotometer® NP80 UV/Vis spectrophotometer for NanoVolume and standard cuvette applications colored touch screen scan range 200 - 900 nm detection range 1 - 16,500 ng/µl dsDNA 1 * 1 ST
Catalog # IMPLNP80TOUCH
Supplier:  IMPLEN
Implen NanoPhotometer® NP80 UV/Vis spectrophotometer for NanoVolume and standard cuvette applications colored touch screen scan range 200 - 900 nm detection range 1 - 16,500 ng/µl dsDNA 1 * 1 ST
Catalog # IMPLNP80TOUCH
Supplier:  IMPLEN