Order Entry
China
ContactUsLinkComponent
MIH-DX including: Stainless steel enclosure Two (2) 20 watt fluorescent lamps with electronic ballasts, non-flickering, independently controlled Adjustable position angle, non-glare black/white background Courtesy side shields Acoustical foam lined i 1 * 1 items
  IBSAMIH-DX
MIH-DX including: Stainless steel enclosure Two (2) 20 watt fluorescent lamps with electronic ballasts, non-flickering, independently controlled Adjustable position angle, non-glare black/white background Courtesy side shields Acoustical foam lined i 1 * 1 items
  IBSAMIH-DX