Order Entry
China
ContactUsLinkComponent
Wash bottles, Capacity 1 Liter. Narrow-mouth bottles, made from PE-LD coloured blue. Screw cap and spray tube from PP natural. Thread GL 32. Ø mm 92. Height mm 221. 1 * 5 items
  VITL132808
 :  VITLAB
Wash bottles, Capacity 1 Liter. Narrow-mouth bottles, made from PE-LD coloured blue. Screw cap and spray tube from PP natural. Thread GL 32. Ø mm 92. Height mm 221. 1 * 5 items
  VITL132808
 :  VITLAB